Learn More
KleenGuard™ Fluidshield™ Procedure Masks
Catalog No. 189992883 Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
Fastening Type:
Earloops
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Fastening Type
18-999-2883 Blue Earloops
18-999-4799 - Earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-2883 Supplier KleenGuard™ Supplier No. 47080
This item is not returnable. View return policy
Face masks are constructed of fluid resistant materials and are lighter and cooler than average procedure masks.
  • Three-layer, pleat-style mask
  • Ear loops
  • Inner facing is light and breathable to reduce irritation
  • Outer facing resists fluid strike through
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Construction 3-ply
Filter Efficiency ≥97% (PFE), ≥96% (BFE)
Latex Free Yes
Resistance 160 mm Hg
Series Fluidshield™