Learn More
Ansell KleenGuard™ V60 30-06™ Readers Safety Eyewear
GREENER_CHOICE
Catalog No. 191301934 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Tortoise Shell
Lens Tint:
Brown
Clear
Yellow
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Magnification Power
19-130-1934 Black Yellow +2.00
19-170-005 Tortoise Shell Brown +2.50
19-170-002 Black Clear +2.50
19-706-016 Black Clear +3.00
19-706-012 Black Clear +1.00
19-130-1933 Black Yellow +1.50
19-130-1935 Black Yellow +2.50
19-170-004 Tortoise Shell Brown +2.00
19-170-003 Tortoise Shell Brown +1.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-130-1934 Supplier Ansell Supplier No. 19894
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard V60 30-06 Readers Safety Glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
  • Soft, comfortable self-adjusting rubber nose pad
  • Precise placement of lens inserts
  • 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Yellow
Product Type Safety Readers
Frame Color Black
Magnification Power +2.00
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400