Learn More
Ansell KleenGuard™ V60 30-06™ Readers Safety Eyewear
GREENER_CHOICE
Catalog No. 191300920 Shop All Ansell Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-130-0920 +2.00
19-130-0919 +1.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-0920 Supplier Ansell Supplier No. 19879
  • $59.40
    $34.65 / Each
    Save $24.75
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $309.50
    $218.65 / Case of 6 EA
    Save $90.85
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard V60 30-06 Readers Safety Glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
  • Stylish ergonomic black nylon frame
  • Sleek seven-base clear polycarbonate lenses
  • Soft rubber nose pad with self-adjusting fingers
  • Magnification insert is molded into the lens
  • UV protection up to 400 nanometers
Compliance
Exceeds ANSI Z87.1-2010 High Velocity Industrial Protection Standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Product Type Prescription Safety Eyewear
Frame Color Black
Magnification Power +2.00
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400