Learn More
Ansell KleenGuard™ V80 MonoGoggle™ Safety Goggles
Catalog No. 172771A Shop All Ansell Products
Change view
Click to view available options
Product Type:
Comfort foam frame
Respirator Fit
Smoke frame
Standard frame
Standard frame; Indirect vent
Lens Coating:
Anti-fog
Anti-fog, UV-treated
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Lens Coating
17-277-1A Standard frame Anti-fog
17-277-1B Respirator Fit Anti-fog, UV-treated
17-277A Standard frame; Indirect vent Anti-fog, UV-treated
19-062307 Comfort foam frame Anti-fog, UV-treated
19-706-025 Smoke frame Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-277-1A Supplier Ansell Supplier No. 14387
This item is not returnable. View return policy
Protect your team and yourself from splashes with the KleenGuard V80 Monogoggle. The safety splash goggles give you eye protection from liquids with a two-piece frame which includes a separate soft, cushioning nose piece for enhanced comfort and fit.
  • Standard or fog-resistant VisiClear™ polycarbonate lens provides 99.9% UV protection up to 380nm
  • Patented ventilation system minimizes condensation while internal baffles protect eyes from dust and chemical splash
  • Soft, pliable nosepiece molds to facial contours
  • Headband attaches at the back of the frame for full peripheral vision
  • Standard MonoGoggle with VisiClear with standard PVC frame or with foam-padded frame for additional impact resistance and comfort
  • Respirator-Fit MonoGoggle designed to be worn with a half-mask cartridge or disposable respirator, without inhibiting their protective fit
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Type VisiClear Lens
Certifications/Compliance ANSI Z87.1-2010
Description Fog-resistant VisiClear; Indirect vent
Lens Tint VisiClear Lens
Product Type Standard frame
For Use With (Application) Safety
Includes Standard Headband Frame
Material Polycarbonate
Frame Material PVC