Learn More
Ansell KleenGuard™ V80 Revolution™ OTG Safety Goggles
GREENER_CHOICE
Catalog No. 191300480 Shop All Ansell Products
Change view
Click to view available options
Certifications/Compliance:
ANSI Z87.1-2010
Catalog No. Certifications/Compliance
19-130-0480 ANSI Z87.1-2010
1 options
Catalog No. 19-130-0480 Supplier Ansell Supplier No. 14399
Add to Cart
Add to Cart
When you need splash protection, anti-fog lenses and a sleek look rolled into one, trust KleenGuard V80 Revolution OTG Safety Goggles.
  • Impact-resistant polycarbonate lenses with VisiClear™ antifog, -scratch and -static coating
  • Easy-to-clean material conforms to face
  • Cross Vent™ Technology keeps air flowing in and moisture or chemical splashes out
  • Blue frames with headband
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Type VisiClear
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Product Type Safety Goggles
Frame Color Blue
Ventilation Details Cross vent
Material Polycarbonate