Learn More
Ansell KleenGuard™ V80 SG34 Safety Goggles
Catalog No. 1798683N Shop All Ansell Products
Change view
Click to view available options
Description:
Anti-fog Lens
Standard lens
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
17-986-83N Standard lens
17-986-83R Anti-fog Lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-986-83N Supplier Ansell Supplier No. 16674
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
When you need comfortable splash protection goggles at an economical price point, look no further than the KleenGuard V80 SG34 Safety Goggles.
  • Polycarbonate lens
  • Button vents prevent indirect venting
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1-2010
Description Standard lens
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Green
Material Polycarbonate