Learn More
Ansell KleenGuard™ V90 Shield™ Safety Goggles with Face Shield
Catalog No. 19706028 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Clear
Catalog No. Lens Tint
19-706-028 Clear
1 options
This item is not returnable. View return policy
Catalog No. 19-706-028 Supplier Ansell Supplier No. 18629
  • $157.80
    $90.65 / Each
    Save $67.15
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $823.00
    $495.65 / Case of 6 EA
    Save $327.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
When you have a job to tackle that involves flying particles or splashes, wearing safety glasses isn't enough. Protect yourself with the KleenGuard V90 Shield Safety Goggles with Face Shield.
  • MonoGoggle XTR with full-face protection
  • Shield detaches from goggle
  • Blue frame with Visiclear Clear lens
  • Replacement shield available (see cat. no. 172831)
Compliance
Meets ANSI Z87.1+ 2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Provides full eye and face protection from liquids without primary eye protection underneath a face shield
Frame Color Blue
Material Polycarbonate
UV Protection UV400