Learn More
Kimtech™ A5 Sterile Cleanroom Boots, Grasp Ties and Vinyl Edge
BIOPHARMAGREENER_CHOICE
Catalog No. p-4639330 Shop All Ansell Products
Change view
Click to view available options
Size:
Small/Medium
Universal
X-Large/2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-902 Small/Medium
19-169-903 Universal
19-169-904 X-Large/2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-169-902 Supplier Kimtech™ Supplier No. 31683
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Sterile cleanroom boots with a wrap-around vinyl foot feature innovative CLEAN-DON™ technology for aseptic donning. Kimtech™ A5 Sterile Cleanroom Boots with Grasp Ties and Vinyl Edge suitable for EU GMP ISO 5 Grade A sterile cleanrooms.
  • Sterile cleanroom boots with grasp ties and vinyl edge
  • Average bacterial filtration efficiency (BFE) of 96%
  • Made of 100% polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-Don ties and 2-inch edge vinyl, for an enhance moisture barrier
  • Made without latex, silicone or the BHT preservative and are designed for single-use
  • 100 Pairs/200 Each
  • CE certified (CE Simple)
  • Each individually bagged pair of boot covers has a sterilization indicator as a visual that it's undergone gamma sterilization
  • A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
TRUSTED_SUSTAINABILITY
Specifications
Size Small/Medium
Material SMS
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Side-grasp Ties
Sole Type Non-skid
Packaging Quantity 100 Pairs/Cs.
Packaging Type Triple Bagged, Vacuum Packed
Resistance None
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.