Learn More
Ansell Kimtech™ A7 Cleanroom Lab Coats
BIOPHARMAGREENER_CHOICE
Catalog No. 19169108 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
4X-Large
6X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-108 Large
19-169-106 Small
19-169-107 Medium
19-169-109 X-Large
19-169-110 2X-Large
19-169-111 4X-Large
19-169-112 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-169-108 Supplier Ansell Supplier No. 47653
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lab coats provide protection and prevent contamination from liquid splashes in controlled environments.
  • Available in bulk cleanroom packaging (double bag)
  • Anti-static treatment and are low lint to reduce contamination risk
  • High collar for increased neck protection
  • Comfortable thumb loops to maintain glove and garment interface
  • Disposable lab coats are recyclable through the RightCycle Program
  • Enhanced chemical and biological protection (passes ASTM F1670 for penetration of blood and body fluids)
  • Protection for people and processes against liquid splashes
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Size Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Static
Collar Type High Mandarin Collar
Latex Free Yes
Seam Type Serged, Triple Overlock Stitching
Certifications/Compliance ASTM F1670
Certifications/Compliance (Europe) EN 1149-5:2018 Type 6 Limited Splash, EN 1149-1:1995 - Antistatic; CE 0120
Includes Thumb Loop
Packaging Type Bulk, Double Bagged