Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A20 Blue Coveralls, Attached Hood and Boots
GREENER_CHOICE

Catalog No. 19057401B Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-057-401B Large
19-057-401C X-Large
19-057-401E 3X-Large
19-057-401D 2X-Large
19-057-520 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-057-401B Supplier Ansell Supplier No. 58523
Add to Cart
Add to Cart
This item is not returnable. View return policy

Denim blue color coveralls feature a breathable and abrasion-resistant fabric to keep workers cool and comfortable while protecting against fibers, fine dusts, and low surface tension liquid splashes.

  • Patented Reflex™ design is 7.5 times less likely to rip out than ANSI minimums.
  • Provides 12% more chest room and 6% longer body length.
  • Ideal for use in dusty environments, as well as for painting, manufacture spraying and agricultural spraying.
  • Stylish denim blue color masks dirt and grease.
  • Air and moisture vapor can pass through to aid wearer comfort.
  • Triple-stitched serged seams.
  • Can withstand multiple wearing.
  • Zippered front, elastic back, attached hood and boots.
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Fabric
Color Denim Blue
Disposable Limited-use
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Dust
Back Style Elastic
Certifications/Compliance NFPA 99
Description Microforce™ Barrier SMS Fabric
Model KleenGuard™ A20
For Use With (Application) Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.