Learn More
Ansell KleenGuard™ A20 Breathable Particle Protection Hood
GREENER_CHOICE
Catalog No. 013498 Shop All Ansell Products
Change view
Click to view available options
Certifications/Compliance:
NFPA 99
Size:
Universal
Catalog No. Certifications/Compliance Size
01-349-8 NFPA 99 Universal
1 options
This item is not returnable. View return policy
Catalog No. 01-349-8 Supplier Ansell Supplier No. 36890
  • $647.00
    $349.65 / Case of 100
    Save $297.35
    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured to the highest standards. KleenGuard™ A20 Breathable Particle Protection Hood offers superior protection.
  • Patented Microforce™ Barrier SMS Fabric
  • Hood extends over shoulders to fit inside coveralls
  • Elastic face opening
  • White
TRUSTED_SUSTAINABILITY
Specifications
Product Type Open Face
Size Universal
Certifications/Compliance NFPA 99