Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A20 Lab Coat with Knit Cuffs

Catalog No. 19999977 Shop All Ansell Products
Change view
Click to view available options
Length (English):
38 in.
44 in.
Length (Metric):
97cm
112 cm
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Size
19-999-977 44 in. 112 cm X-Large
19-999-974 38 in. 97cm Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-999-977 Supplier Ansell Supplier No. 23874

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Breathable particle protection with comfortable snug fit

Jackets feature snug-fitting polyester-knit cuffs on raglan sleeves.

  • With high mandarin collar, one chest pocket and one hip pocket
  • Plastic snap-front closure
  • Denim blue

TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material SMS
Color Denim Blue
Closure Type Snaps
No. of Pockets 2
Collar Type High mandarin
Certifications/Compliance NFPA 99
For Use With (Application) Suited for laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
Length (English) 44 in.
Length (Metric) 112 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.