Learn More
Ansell KleenGuard™ A40 Liquid and Particle Protection Coveralls
Catalog No. 191303477E Shop All Ansell Products
Change view
Click to view available options
Zipper front; Individually wrapped To-Go; XX-Large
Catalog No.
19-130-3477E Zipper front; Individually wrapped To-Go; XX-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-130-3477E Supplier Ansell Supplier No. 44345
This item is not returnable. View return policy
Excellent protection from nonhazardous liquids
Microporous, nonwoven film laminate offers an improved liquid barrier over SMS apparel.
  • Low linting
  • White
  • Zipper front
Compliance
Meets ANSI™/ISEA™ 101-1996; ASTM™ F1670 testing for penetration of blood and bodily fluids; NFPA 99 Health Care Facilities criteria for antistatic materials.
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material SMS
Color White
Disposable Single-use
Closure Type Front Zipper
Certifications/Compliance ASTM F1670, NFPA 99, ANSI ISEA 101-1996
Description Zipper front; Individually wrapped To-Go; Non-woven Microporous film laminate barrier over SMS
For Use With (Application) Fiberglass manufacturing, liquid handling, utilities, pressure washing, tank cleaning, maintenance and general manufacturing
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.