Learn More
Kimberly-Clark Professional™ KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
Manufacturer:  Kimberly-Clark Professional™ 45002
 View more versions of this product
Catalog No. 19-057-403A
Add to cart
Description
Description
Breathable fabric is strong, comfortable, and abrasion-resistant, with a three-layer construction designed to allow heat and sweat vapor to escape.
  • Patented Reflex™ Design is 7.5 times less likely to rip out than ANSI™ minimum standards
  • Feature 12% more chest room and 6% longer body length
  • Provide excellent front and strikethrough protection with a seamless front and concealed 28 in. (71cm) zipper
  • Used in many industries and applications, including emergency medical, liquid handling, forensics, nuclear facilities and more
  • Denim blue
Compliance
Meet ASTM F1670/1671 specifications for blood, bodily fluids and bloodborne pathogens.
Specifications
Specifications
Elastic
Elastic
Zipper with Storm Flap
Elastic
Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing
Medium
Safety
ASTM F1670, 1671
Denim Blue
Elastic wrist and ankle, seamless zipper with storm flap, elastic across the back, Reflex™ design; 3-ply Fabric construction middle layer of Microporous film
Fabric
Documents
Accessories