Learn More
Ansell KleenGuard™ A70 Chemical Splash Protection Sleeves
Catalog No. 19166519 Shop All Ansell Products
Change view
Click to view available options
Material:
Composite Fabric
Catalog No. Material
19-166-519 Composite Fabric
1 options
This item is not returnable. View return policy
Catalog No. 19-166-519 Supplier Ansell Supplier No. 97780
  • PRODUCT PROMOTION
    $1,002.00
    $445.42 / Case of 200
    Save $556.58
    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Resistant to many water-based liquids, greases and oils, and liquid chemical sprays
  • Yellow polyethylene-coated fabric
  • Elastic cuffs
  • 21.25 in. long (54cm)
TRUSTED_SUSTAINABILITY
Specifications
Material Composite Fabric
Cuff Style Elastic