Learn More
Ansell Smith & Wesson™ 44 Magnum™ Safety Eyewear
Catalog No. 19706023 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Lens Tint:
Clear
Indoor/Outdoor
Smoke
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-706-023 Black Indoor/Outdoor
19-706-021 Black Smoke
19-706-022 Black Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-706-023 Supplier Ansell Supplier No. 23454
Add to Cart
Add to Cart
This item is not returnable. View return policy
Scratch-resistant, antifog polycarbonate lens provides 99.9% UV protection against harmful UV rays
  • Designed with ergonomic features for long-term performance and all-day comfort
  • Soft-touch temples naturally contour to the head for a secure and comfortable fit
  • Cut-to-shape distinctive and stylish one-piece lens provides front and side protection
  • Vents on temples maximize airflow and minimize fogging
  • Flexible nosepiece conforms to various nose shapes; provides protection by minimizing gap between face and lens
Compliance
Meets ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Indoor/Outdoor
Frame Color Black
Lens Material Polycarbonate