Learn More
Ansell Smith & Wesson™ Magnum 3G™ Eyewear
Catalog No. 191300902 Shop All Ansell Products
Change view
Click to view available options
Size:
Full-sized
Mini
Frame Color:
Black
Metallic Gray
Platinum
Lens Tint:
Clear
GIO
Smoke
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Frame Color Lens Tint
19-130-0902 Mini Black Smoke
19-806-501 Mini Black Clear
19-150-2000 Full-sized Platinum Clear
19-804-400 Full-sized Black Smoke
19-804-280 Full-sized Metallic Gray GIO
19-806-500 Full-sized Black Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-0902 Supplier Ansell Supplier No. 19824
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ergonomically dimensioned three-point fit for comfort
  • Built-in hinge stops help to eliminate scratches
  • Self-adjusting ergonomic rubber nose pad with rubber fingers
  • Mini-Magnum™ designed to fit small faces
  • Provides 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Smoke
For Use With (Application) Magnum Mini - reduced size for smaller faces
Frame Color Black
Includes Self-adjusting nose pad w/rubber fingers
Size Mini