Learn More
Ansell Smith & Wesson™ Magnum 3G™ Eyewear
Ergonomically dimensioned three-point fit for comfort
Supplier:  Ansell 19823
 View more versions of this product
Catalog No. 19-804-400
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Ergonomically dimensioned three-point fit for comfort
  • Built-in hinge stops help to eliminate scratches
  • Self-adjusting ergonomic rubber nose pad with rubber fingers
  • Mini-Magnum™ designed to fit small faces
  • Provides 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Meets ANSI Z87.1-2003 standards.
Smoke
Black
Full-sized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.