Learn More
Kimberly-Clark Professional™ Smith & Wesson™ Phantom Safety Eyewear
Click to view available options
Lens Tint:
Clear anti-fog lens
Clear lens
Smoke Tint Lens
Frame Color:
Black
Pewter
Description
- Seven-base spherical lens
- Ribbed, non-slip rubber nose pad
- Straight-back bayonet design temples eliminate mastoid bone pressure
- UV protection
Compliance
Meets ANSI Z87.1 - 2010 High Velocity Industrial Standards
Specifications
Certifications/Compliance | Exceeds ANSI Z87.1 - 2010 High Velocity Industrial Standards |
Lens Tint | Smoke Tint Lens |
Frame Color | Black |
Temple Style | Straight back bayonet design |