Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell X-Tra™ Traction Shoe Covers
GREENER_CHOICE

Catalog No. 190202501 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium/Large
X-Large/2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-2501 Medium/Large
19-020-2502 X-Large/2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-020-2501 Supplier Ansell Supplier No. 69252
  • $608.57
    $494.65 / Case of 300

    Save $113.92

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Shoe covers made with SMS fabric feature adhesive traction strips on the bottom for stability.

  • Light blue, three-layer SMS fabric
  • Fully elasticized
  • Adhesive strips for traction on bottom
TRUSTED_SUSTAINABILITY

Specifications

Size Medium/Large
Material SMS
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Traction Strips
Model 69252
Includes Traction
For Use With (Application) Laboratories, Healthcare
Packaging Quantity 300/Cs.
Product Type Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.