Learn More
Ansell Safeskin™ Critical Nitrile Gloves
BIOPHARMAGREENER_CHOICE
Catalog No. 19059501 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-059-501 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-059-501 Supplier Ansell Supplier No. 56884
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safeskin™ critical nitrile gloves feature textured fingertips to facilitate control and grip.
  • Clean processed for low particle and extractable levels
  • Ambidextrous with beaded cuff for added strength
  • Textured fingertips for improved grip
  • Silicone-free formulation
  • Double-lined bags
TRUSTED_SUSTAINABILITY
Specifications
Color White
Exterior Finish Partially Textured
Hand Ambidextrous
Material Nitrile
Powdered Powder Free
Size Large
Sterility Non-sterile
Thickness (English) 5.1 mil
Length (English) 12 in.
Length (Metric) 30.5 cm