Learn More
Ansell Smith & Wesson™ Elite Safety Glasses
Catalog No. 19169894 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Indoor/Outdoor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-169-894 Amber
19-169-895 Indoor/Outdoor
19-169-893 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-169-894 Supplier Ansell Supplier No. 21303
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Elite Safety Glasses are designed to be both stylish and comfortable, with a range of options for workers during both work and play
  • Lightweight
  • Retro design that provides a stylish fashion statement
  • Wide temples provide structural integrity and stability
  • Designed for all-day comfort whether on the job, at the gun range, doing yard work, or driving
  • Provide 99.9% UVA/UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Anti-scratch, dual lens offer wrap-around protection
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Amber
Frame Color Black
Includes Micro-fiber carry pouch
No. per Case 12/Cs.
Temple Style Wide