Learn More
Ansell Smith & Wesson™ Equalizer™ Safety Glasses
Catalog No. 19169887 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor
Smoke
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-169-887 Smoke Anti-fog, Scratch-resistant
19-169-888 Indoor/Outdoor Scratch-resistant
19-169-876 Clear Anti-fog, Scratch-resistant
19-169-875 Clear Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-169-887 Supplier Ansell Supplier No. 21297
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Equalizer™ Safety Glasses feature stylized frame with sport-grip temples to help reduce slippage while the dual lenses offer wraparound protection
  • 99.9% UV (UVA / UVB / UVC) protection and meet the high impact level for the ANSI Z87.1+ standards for personal eye protection
  • Each pair comes with a microfiber carry pouch that doubles as a cleaning cloth
  • Polycarbonate lenses help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Anti-scratch coating on every lens to prevent lens damage
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Smoke
Frame Color Gun Metal
Includes Micro-fiber carry pouch
No. per Case 12/Cs.
Temple Style Sport Grip
Lens Material Polycarbonate