Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Soft Touch™ II Surgical Mask

Catalog No. 189994819 Shop All Ansell Products
Change view
Click to view available options
Surgical mask; Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-999-4819 Surgical mask; Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4819 Supplier Ansell Supplier No. 47500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Three-layer construction protects nose and mouth with comfortable fit.

  • Pleat style
  • Horizontal ties
  • Blue
  • One-size-fits-all

Applications:

Ideal for surgical and dental use

TRUSTED_SUSTAINABILITY

Specifications

Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Ties
Material Fabric
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-ply
Filter Efficiency ≥97% (PFE), ≥96% (BFE)
Includes Horizontal ties
Model 47500
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Soft Touch II Surgical Mask
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.