Learn More
Ansell Standard Cone-Style Surgical Mask
Catalog No. 19166977 Shop All Ansell Products
Change view
Click to view available options
:
Headgear with faceshield
Catalog No.
19-166-977 Headgear with faceshield
1 options
This item is not returnable. View return policy
Catalog No. 19-166-977 Supplier Ansell Supplier No. 00152
This item is not returnable. View return policy
Standard Cone-Style Surgical Mask is appropriate for situations where exposure to blood and/or bodily fluids is not a risk
  • Materials and donning attachments are sonically bonded
  • Mask is natural rubber latex-free
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Color Blue
Fastening Type Headband
Material Polyester
Size Standard
Style Cone
Construction 3-ply
Filter Efficiency 0.95
Model 152
Pore Size 3 μm (BFE)