Learn More
Ansell Sterile Evolution™ 4 Surgical Gowns
Catalog No. 19810267 Shop All Ansell Products
Change view
Click to view available options
Material:
SMS
Size:
X-Large
Sleeve Style:
Set-in Sleeve
Catalog No. Material Size Sleeve Style
19-810-267 SMS X-Large Set-in Sleeve
1 options
This item is not returnable. View return policy
Catalog No. 19-810-267 Supplier Ansell Supplier No. 90048
This item is not returnable. View return policy
Fourth-generation fabric technology provides advanced protection, breathability and comfort
  • Evolution 4 SMS fabric
  • Adjustable neckline with yellow neck binding - hook and loop
  • Resistant to fluid, flame and lint
  • Meets NFPA Class I flame resistance
  • Abrasion resistant
  • Sterile
Set-in Sleeves
  • Non-reinforced fabric
  • Book fold
Raglan Sleeves
  • Reinforced fabric
  • Drop fold
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material SMS
Color Blue
Closure Type Snaps
Disposable Single-use
Sterility Sterile
Model 90048
Sleeve Style Set-in Sleeve
Resistance Fluid, flame and lint-resistant
Certifications/Compliance AORN
Autoclavable Not Autoclavable