Description
- Four-point ratchet temples to provide a customized fit for long wear comfort
- Brow guard for enhanced coverage and protection
- Meets ANSI Z87.1+ standards for impact and is CSA certified to meet the requirements of CSA Z94.3
- Polycarbonate lenses are hardcoated for scratch protection and longer life and provide 99.9% UVA / UVB / UVC
Specifications
Specifications
| Lens Tint | Smoke |
| Frame Color | Black |
| No. per Pack | 12/Pk. |
| Temple Style | Adjustable ratchet |