Learn More
Ansell V40 HellRaiser™ Safety Glasses
Catalog No. 19169871 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Amber
Blue
Blue Mirror
Clear
IR Filter Shade 3.0
IR Filter Shade 5.0
Indoor/Outdoor
Smoke
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-169-871 Clear Anti-fog, Scratch-resistant
19-800-002 Indoor/Outdoor Scratch-resistant
19-800-000 Clear Scratch-resistant
19-800-001 Smoke Scratch-resistant
19-800-005 Blue Mirror Scratch-resistant
19-800-007 IR Filter Shade 5.0 Scratch-resistant
19-800-004 Blue Scratch-resistant
19-800-006 IR Filter Shade 3.0 Scratch-resistant
19-800-003 Amber Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-169-871 Supplier Ansell Supplier No. 28615
Add to Cart
Add to Cart
This item is not returnable. View return policy
V40 HellRaiser™ Safety Glasses feature a high-performance, durable polycarbonate lenses, which provide 99.9% UV protection (UVA/UVB/UVC)
  • Lightweight and stylish with single lens wraparound protection designed for use at work or home
  • Flex-point temples help provide comfort and stability for use for long stretches of time
  • Ideal for industrial manufacturin
  • Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
  • Hard coated lenses for scratch resistance and longer life
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Frame Color Black
No. per Case 12/Cs.
Temple Style Flex point
Lens Material Polycarbonate