Learn More
DWK Life Sciences KIMBLE™ French Square Glass Bottles
SDP
Catalog No. 13756373BLK Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Soda Lime Glass
Catalog No. Material
13-756-373BLK Soda Lime Glass
1 options
Catalog No. 13-756-373BLK Supplier DWK Life Sciences Supplier No. 30772900999
Add to Cart
Add to Cart
KIMBLE™ French Square Glass Bottles are made from clear USP Type III soda-lime glass. The wide-mouth, square design maximizes shelf storage space and is ideal for solvent, chemical, or sample storage. Available with caps attached or in bulk packs without caps.
  • Clear USP Type III soda-lime glass construction
  • Square shape maximizes use of shelf and storage space
  • Wide-mouth design for efficient addition and removal of contents
  • Ideal for solvent, chemical, or sample storage
  • Available with caps attached or in bulk packs without caps
  • 48-400 GPI finish
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 473 mL
Color Clear
Material Soda Lime Glass
Closure Included No
Graduated No
Mouth Wide
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape French Square
Product Type French Square Bottle