Learn More
DWK Life Sciences™ Kimble™ KIMAX™ Distilling Apparatus Complete Assembly
Complete assembly
$1009.57
Specifications
Type | Distillation Apparatus |
---|---|
Material | Borosilicate Glass |
Certifications/Compliance | USP Type I and ASTM E438, Type I, Class A |
Standard Taper Joint | 19/38 |
Description
Use to distill small amounts of water or in APHA Method 4500-NH3 for determining nitrogen (ammonia) in purified drinking water, natural water and highly purified wastewaters (concentration, <20μg/L)
Specifications
Distillation Apparatus | |
Borosilicate Glass | |
USP Type I and ASTM E438, Type I, Class A | |
19/38 |