Learn More
DWK Life Sciences Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Extraction tube only
SDP
For use with for Kimble™ extraction apparatus
$587.71 - $756.32
Specifications
Product Type Extraction Tube
For Use With (Equipment) Soxhlet Extraction Apparatus
Standard Taper Joint 24/40
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (English) Inside Diameter (Metric) Inside Standard Taper Joint Top Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inside Diameter (Metric) Inside Standard Taper Joint Top Price Quantity & Availability  
09-553A
SDP
View Documents
DWK Life Sciences
2407130
1.18 in. 2.96 cm 34/45
Case for $587.71
Only null left
Add to Cart
09-553B
SDP
View Documents
DWK Life Sciences
2407140
1.57 in. 4 cm 45/50
Case for $663.04
Only null left
Add to Cart
09-553C
SDP
View Documents
DWK Life Sciences
2407150
1.96 in. 5 cm 55/50
Case for $756.32
Only null left
Add to Cart
Description
Description
  • With standard taper 24/40 joint at bottom
  • Connect by interchangeable ground joints
Specifications
Specifications
Extraction Tube
Soxhlet Extraction Apparatus
24/40
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.