Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ KIMBLE™ Replacement Snap Cap
GSA_VASDP

Catalog No. 03340720 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
03-340-720 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-340-720 Supplier DWK LIFE SCIENCES™ Supplier No. 738372
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ Replacement Snap Cap is a polyethylene closure designed for use with KIMBLE™ centrifuge tubes (Cat. No. 05-538-41C/D). Provides a secure, reliable fit for compatible tube formats.

  • Polyethylene construction for chemical compatibility and durability
  • Size 2 snap cap designed as a replacement closure
  • Compatible with Kimble centrifuge tubes Cat. No. 05-538-41C/D

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Snap Cap
Autoclavable Not Autoclavable
Material Polyethylene
Color White
Sterility Non-sterile
For Use With (Equipment) 05-538-41C/D Disposable Plain Conical Glass Centrifuge Tubes
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.