Learn More
DWK Life Sciences Kimble™ Kontes™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection
SDP
Catalog No. K9537600000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
125 mL
1000 mL
2000 mL
4000 mL
Standard Taper Joint:
5
8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
K953760-0000 1000 mL 8
K953710-0000 125 mL 5
K953760-2000 2000 mL 8
K953760-4000 4000 mL 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K953760-0000 Supplier DWK Life Sciences Supplier No. 9537600000
Add to Cart
Add to Cart
Borosilicate glass flask with side arm, No. 2 hose connection.
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection, Connector Types: No. 2 Hose Connection, Borosilicate Glass, Description: Filtration Flask
Order Info
Does not include rubber stopper and hose
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Includes No. 2 Hose Connection
Product Type Flask with Side Arm
Standard Taper Joint 8
Quantity 1/Cs.