Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ HPLC Solvent Pickup Ultra-Ware™ Adapters: THF-Resistant Pickup Adapter, 90mm
SDP

Catalog No. K9539066390 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inside:
90 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inside
K953906-6390 90 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K953906-6390 Supplier DWK Life Sciences Supplier No. 9539066390
Add to Cart
Add to Cart

For use with five valve, four valve and filtration caps

  • Provides safe, in-line filtration that replaces the traditional pour-and-wait funnel filtration method
  • Unique Bevel-Seal makes a vacuum tight o-ring connection to the PTFE tubing
  • Hi-Vac valve is easier to open under vacuum than the standard stopcock
  • Closing the valve provides vacuum degassing after filtration
  • Each unit is supplied with 3 ft. (91cm) of 1/4 O.D. FEP PTFE tubing
  • GL-45 solvent bottle adapter is made from PBT and converts Ultra-ware GL-45 mobile phase caps to standard 4L solvent bottles for direct access to HPLC instruments

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Five valve, Four valve and filtration caps
Material PBT
Type THF-Resistant Pickup Adapter
Diameter (Metric) Inside 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.