Learn More
DWK Life Sciences Kimble™ Kontes™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
GSA_VASDP
Catalog No. p-73275 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
13-415
15-415
24-400
28-400
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03-340-65 24-400
03-340-70B 28-400
03-340-77AA 13-415
03-340-77BB 15-415
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-340-65 Supplier DWK Life Sciences Supplier No. 7380824400
Only null left
Add to Cart
Add to Cart
For use with screw-thread culture and centrifuge tubes
  • Provides superior closure system capable of maintaining its liner through multiple autoclavable operations
  • PTFE-coated surface of exposed liner is chemical-resistant and compatible with sensitive, corrosive and volatile solvents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Screw Cap
Closure Size 24-400
Liner Material PTFE, Rubber
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) For repeated autoclaving
Tip Material PTFE
For Use With (Equipment) EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes