Learn More
DWK Life Sciences Kimble Chase™ Tall-Form High Precision Specific Gravity Hydrometers
Available in a variety of graduation ranges and overall lengths
Supplier: DWK Life Sciences 521111414
Description
- Serial number printed on the scale
- Ballast is made from steel pellets and a binder, and is free of heavier metals
- Includes Certificate of Traceability
- Certificate of Traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
- Meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
Specifications
Tall-Form High Precision Specific Gravity Hydrometer | |
330 mm | |
7 mm | |
1.360 to 1.430 |
0.0005 sp.gr. | |
Certificate of Traceability | |
21 mm | |
600 mL |