Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ GLS 80™ KimCote™ Clear Wide Mouth Media Bottles with Caps
SDP

Catalog No. 02913082 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
5000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-913-082 500 mL
02-913-081 250 mL
02-913-083 1000 mL
02-913-084 2000 mL
02-913-085 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-913-082 Supplier DWK Life Sciences Supplier No. 143900500
Add to Cart
Add to Cart

Get versatility, purity and durability to achieve results for laboratory storage, bioprocess and production applications.

  • Wide-mouth
  • 3.3 borosilicate glass
  • Durable ceramic ink graduations and marking patch
  • Blue quick-release linerless polypropylene closure and pour ring are rated up to 140° C
  • Autoclavable
  • KimCote™ coating provides safety and reduces hazards of shattered glass
Note: Image is representative only. Bottle capacity may vary.

TRUSTED_SUSTAINABILITY

Specifications

Bottle Surface KimCote™
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Coating KimCote™
Color Clear
For Use With (Application) Storage and Transport
Includes Blue Polypropylene Cap
Material Borosilicate Glass (3.3)
Length (Metric) 225.425 mm
Max. Temperature (Metric) 140°C
Packaging Case
Quantity 4 Pk.
Sterility Non-sterile
Closure Color Blue
Closure Included Yes
Graduated Yes
Height (Metric) 152 mm
Closure Liner Unlined
Closure Material Polypropylene
Closure Size GLS 80
Diameter (Metric) 101 mm
Closure Type Quick-release Screw Thread
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.