Learn More
DWK Life Sciences Kimble™ Kontes™ Center Tear-Off Aluminum Seals, Color Coded
Shop All DWK Life Sciences Products

Description
- Allows total removal of seal without decappers
- For use with 13 and 20mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
Order Info
Available in blue, red, and green color to help separate and identify samples
Specifications
Specifications
| Color | Red |
| Material | Aluminum |
| Quantity | 1000 Cs. |
| Closure Size | 13 mm |
| Product Type | Tear-off |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.