Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Center Tear-Off Aluminum Seals, Color Coded
SDP

Catalog No. 03340322 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
Closure Size:
13 mm
20 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
03-340-322 Red 20 mm
03-340-320 Green 13 mm
03-340-323 Green 20 mm
03-340-321 Blue 20 mm
03-340-319 Red 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 03-340-322 Supplier DWK Life Sciences Supplier No. 73821C20
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tamper-evident center tear-off feature protects target area

  • Allows total removal of seal without decappers
  • For use with 13 and 20mm serum finished bottles and vials
  • Designed to secure septa and stoppers to bottles and vials

Order Info

Available in blue, red, and green color to help separate and identify samples

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Aluminum
Quantity 1000 Cs.
Closure Size 20 mm
Product Type Tear-off Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.