Learn More
DWK Life Sciences Kimble™ Kontes™ Friedrich Condensers
SDP
Catalog No. 10310234 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
24/40
Catalog No. Standard Taper Joint No
10-310-234 24/40
1 options
Catalog No. 10-310-234 Supplier DWK Life Sciences Supplier No. 3416519
  • Molded inner cold finger
  • Hose connection for 3/8 in. ID flexible tubing
24/40 Joint
  • Specially designed to provide a long vapor path, good heat transfer and anti-flooding characteristics
  • Inclined outer joint on the side and two hose connectors at the top
  • Inner drip joint at the bottom
  • Side arm is an outer standard taper joint and the bottom is an inner standard taper joint
  • Water inlet and outlet located ring-sealed inner condensing tube
  • Inner drip joint and side outlet for drying tube
  • TRUSTED_SUSTAINABILITY
    Specifications
    Material Glass
    Standard Taper Joint No 24/40
    Type Friedrich Condenser