Learn More
DWK Life Sciences Kimble™ Kontes™ Offset BEVEL-SEAL™ Inlet Adapter
SDP
Catalog No. K1797102119 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
19/22
Catalog No. Standard Taper Joint No.
K179710-2119 19/22
1 options
Catalog No. K179710-2119 Supplier DWK Life Sciences Supplier No. 1797102119
Add to Cart
Add to Cart
A unique glass adapter offset and angled approximately 10° for use with multi-neck flasks.
  • Thermometers or bleed tubes are angled into the bottom of a flask
  • A vacuum-tight seal is created with the open top compression cap and FKM o-ring to allow adjustable immersion of thermometers
  • Cap is suitable for use to 150 °C
  • Supplied complete with one size 010 FKM o-ring and one 410119 cap
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Includes One size 010 FKM O-ring and one 410119 cap
Material Borosilicate Glass
Product Type Offset Bevel-Seal™ Inlet Adapter
Standard Taper Joint No. 19/22
Thread Size 13-425