Learn More
DWK Life Sciences Kimble™ Kontes™ Pestle For All-Glass DUALL™ Tissue Grinders
SDP
Catalog No. K8854510021 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
20
21
22
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
K885451-0021 21
K885451-0020 20
K885451-0022 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K885451-0021 Supplier DWK Life Sciences Supplier No. 8854510021
Add to Cart
Add to Cart
Grinding efficiency is greatly improved when this tube is used. Available in sizes 20 - 23.
  • Combines both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate.
  • This ground glass model is ideal for connective tissue such as muscle, heart and lung
  • Construction is strong enough to allow the pestle to be motor-driven
  • Clearance between pestles and tubes is 0.004 in. to 0.006 in.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Size 21
Product Type Tissue Grinder Pestle
For Use With (Equipment) Duall™ all Glass tissue grinders