Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint
SDP

Catalog No. K4225300010 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
125 mL
250 mL
Diameter (Metric) Outer:
2.2 cm
6.5 cm
8.5 cm
Standard Taper Joint:
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint
K422530-0010 10 mL 2.2 cm 19/22
K422530-0250 250 mL 8.5 cm 24/40
K422530-0125 125 mL 6.5 cm 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K422530-0010 Supplier DWK Life Sciences Supplier No. 4225300010

Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids

  • Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
  • Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
  • Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
  • Adjust spray pattern by varying the position of thumb on vent hole
  • Autoclavable (after cap, loosening ring, and O-ring are removed)

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 0.33 oz.
Capacity (Metric) 10 mL
Connections Screw thread ground joint, connecting cap, loosening ring
Material Glass
Quantity 1 Case
Type TLC Reagent Sprayer
For Use With (Application) To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography
For Use With (Equipment) Standard TLC Reagents and Most Corrosive Liquids
Operating Pressure 3 to 5 psi
Standard Taper Joint 19/22
Height (English) 6.1 in.
Height (Metric) 15.5 cm
Diameter (English) Outer 0.86 in.
Diameter (Metric) Outer 2.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.