Learn More
DWK Life Sciences™ Kimble™ Nickel-Plated, Standard Taper Joint Clamp
This Kimble™ Nickel-Plated metal, Standard Taper Joint Clamp designed to secure Standard Taper joints. The nickel-plating provides increased resistance to heat and cracking.
Supplier: DWK Life Sciences™ 6753200019
This item is not returnable.
View return policy
Description
- Manufactured from Nickel-plated metal
- When using a concentrator tube heating mantle, these clamps may be used to secure the concentrator tube to the concentrator flask
- They can withstand temperatures as high as 200°C, and they have excellent chemical resistance
- Easy and safe to put on and are dimensionally stable
Specifications
Standard Taper Joint Clamp | |
Nickel-Plated Metal | |
19/22, 19/38 |