Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Open-Top Aluminum Seals, Color Coded
SDP

Catalog No. 03340308 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Closure Size:
13 mm
20 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
03-340-308 Green 13 mm
03-340-311 Green 20 mm
03-340-309 Blue 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-340-308 Supplier DWK Life Sciences Supplier No. 73822D13

Designed to secure septa and stoppers to bottles and vials

  • For use with 13 and 20mm serum finished bottles and vials
  • Available in blue, red, green and natural aluminum color to help separate and identify samples

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Aluminum
Quantity 1000 Cs.
Closure Size 13 mm
Product Type Open Top Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.