Learn More
Description
- For use with 13 and 20mm serum finished bottles and vials
- Available in blue, red, green and natural aluminum color to help separate and identify samples
Specifications
Specifications
| Color | Green |
| Material | Aluminum |
| Quantity | 1000 Cs. |
| Closure Size | 20 mm |
| Product Type | Open Top Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
