Learn More
DWK Life Sciences Kimble™ Phenolic Caps with PTFE-Faced LDPE Foam Liners
Shop All DWK Life Sciences Products

Description
- General purpose phenolic screw cap with foam liner
- Color: black
- Liner material: 0.005-in. PTFE-faced 0.050-in. LDPE foam
- Poor for organic amines
- Compressible foam for sealability
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Neck Finish | 24-400 |
| Product Type | Phenolic Cap |
| For Use With (Equipment) | Bottle |
| Closure Liner | PTFE/LDPE Foam |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.