Learn More
Description
- Polyvinyl film is adhered to 0.035-in. pulpboard
- Color: black
- Low cost cap for general purpose usage
- Not recommended for hydrocarbons or bleaches
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Neck Finish | 63-400 |
| Product Type | Phenolic Cap |
| For Use With (Equipment) | Bottle |
| Closure Liner | Pulp/Vinyl |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
