Learn More
Kimtech™ A5 Sterile Integrated Hood and Mask
BIOPHARMAGREENER_CHOICE
Catalog No. 17986053 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-053 X-Large
19-048-606 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-986-053 Supplier Kimtech™ Supplier No. 47677
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleanroom sterile hood with a mask integrated into the hood.
  • Average bacterial filtration efficiency (BFE) of 96%
  • Polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-don ties for easy gowning and are folded inside out for aseptic donning
  • Protective hood-and-mask-combos are made without latex, silicone or the BHT preservative
  • Designed for single-use
  • CE certified, Category III
  • Each individually bagged mask and hood has a sterility indicator
  • Manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
TRUSTED_SUSTAINABILITY
Specifications
Product Type Integrated Hood and Mask
Size X-Large
Material Polypropylene SMS Fabric
Color White
Closure Type Ties
Disposable Single-use
Model 47677
Sterility Sterile
Hood Type Integrated
Cleanroom Class Class 100 (ISO 5)
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.