Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ Element™ Nitrile Gloves
BIOPHARMA

Catalog No. 17000586 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-586 X-Large
17-000-582 X-Small
17-000-583 Small
17-000-584 Medium
17-000-585 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-000-586 Supplier Ansell Supplier No. 62874

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kimtech™ Element™ Nitrile Exam Gloves are reliable, easy-to-wear gloves that are more economical than standard nitrile gloves, and are suited for a wide variety of tasks.

The Kimtech™ Element™ nitrile gloves deliver enhanced dexterity, thanks to textured fingertips, and are not made with natural rubber latex. The gloves are static dissipative in use. They come in an innovative dispenser box, which incorporates two separate openings. The first, smaller opening is used when the box is full and helps control dispensing to lessen waste from dropped gloves or multiple dispensing. When the box is half empty, the second, larger opening allows easier access to the gloves. They have an AQL of 1.5 for pinholes and can even be recycled with the RightCycle program, the first large-scale recycling program for nonhazardous lab and cleanroom waste. Kimtech™ Element™ Nitrile Gloves can be an important piece for creating a more efficient, responsible workplace.

  • Exceptional value: 250% more gloves per case (versus standard nitrile gloves)
  • Not made with natural rubber latex, are powder -free and feature beaded cuffs
  • Superior quality, AQL 1.5 (exceeds the ASTM D 5151 standard for critical defects or pinholes)
  • Waste less with the one-at-a-time dispenser box; Element Nitrile Gloves are recyclable through the RightCycle program
TRUSTED_SUSTAINABILITY

Specifications

Accelerator Free No
Applications Laboratory, Healthcare, Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 3.15 mil
Finger Thickness (Metric) 0.08 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 1.97 mil
Palm Thickness (Metric) 0.05 mm
Powdered Powder Free
Size X-Large
Thickness (English) 2.8 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Length (English) 9 in.
Length (Metric) 22.86 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.